Learn More
Lenz Laborglasintrumente Round-Bottom Flasks With Conical Ground Joint
Product Code. p-7222522
Click to view available options
Capacity (Metric):
500 mL
2 L
3 L
4 L
5 L
6 L
10 L
20 L
Diameter (Metric) Outer:
105 mm
166 mm
185 mm
207 mm
223 mm
236 mm
279 mm
345 mm
Standard Taper Joint:
NS 19/26
NS 29/32
NS 45/40
Unit Size:
Each
Pack of 10
Product Code. 10577311
Brand: Lenz Laborglasintrumente 03004585
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • According to DIN EN ISO 4797
  • With inscription label
  • Material: DURAN™
Specifications
Capacity (Metric) 6 L
Material Duran Glass
Standard Taper Joint NS 45/40
Diameter (Metric) Outer 236 mm
Quantity 1 Ea.