Learn More
Lenz Laborglasintrumente Round-Bottom Flasks With Conical Ground Joint
Click to view available options
Capacity (Metric):
500 mL
2 L
3 L
4 L
5 L
6 L
10 L
20 L
Diameter (Metric) Outer:
105 mm
166 mm
185 mm
207 mm
223 mm
236 mm
279 mm
345 mm
Standard Taper Joint:
NS 19/26
NS 29/32
NS 45/40
Unit Size:
Each
Pack of 10
Description
- According to DIN EN ISO 4797
- With inscription label
- Material: DURAN™
Specifications
Specifications
| Capacity (Metric) | 6 L |
| Material | Duran Glass |
| Standard Taper Joint | NS 29/32 |
| Diameter (Metric) Outer | 236 mm |
| Quantity | 1 Ea. |