Learn More
Lenz Laborglasintrumente Round Bottom Flasks, Narrow Neck
Description
- Acc. to DIN ISO 1773 and in addition to standard
- With beaded rim
- Material: DURAN™
Specifications
Specifications
| Diameter (Metric) Outer Neck | 76 mm |
| Capacity (Metric) | 20 L |
| Height (Metric) | 515 mm |
| Includes | Beaded rim |
| Quantity | 1 Ea. |