Learn More
Lenz Laborglasintrumente Round Bottom Flasks, Narrow Neck
Product Code. 10739221
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
3 L
4 L
5 L
6 L
10 L
20 L
Unit Size:
Each
Pack of 10
Product Code. 10739221
Brand: Lenz Laborglasintrumente 07030085
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN ISO 1773 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Diameter (Metric) Outer Neck 51 mm
Capacity (Metric) 6 L
Height (Metric) 355 mm
Includes Beaded rim
Quantity 1 Ea.