Learn More
Lenz Laborglasintrumente Round Bottom Flasks, Narrow Neck
Product Code. 10507671
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
3 L
4 L
5 L
6 L
10 L
20 L
Unit Size:
Each
Pack of 10
Product Code. 10507671
Brand: Lenz Laborglasintrumente 07030079
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN ISO 1773 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Diameter (Metric) Outer Neck 50 mm
Capacity (Metric) 4 L
Height (Metric) 290 mm
Includes Beaded rim
Quantity 1 Ea.