Learn More
Lenz Laborglasintrumente Round Bottom Flasks, Narrow Neck
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
3 L
4 L
5 L
6 L
10 L
20 L
Unit Size:
Each
Pack of 10
Description
- Acc. to DIN ISO 1773 and in addition to standard
- With beaded rim
- Material: DURAN™
Specifications
Specifications
| Diameter (Metric) Outer Neck | 34 mm |
| Capacity (Metric) | 500 mL |
| Height (Metric) | 168 mm |
| Includes | Beaded rim |
| Quantity | 10/Pk. |