Learn More
Lenz Laborglasintrumente™ Round-Bottom Flasks with Conical Ground Joint
Click to view available options
Capacity (Metric):
250 mL
4000 mL
Unit Size:
Each
Pack of 10
Description
- According to DIN EN ISO 4797 and in addition to standard
- With inscription label
- Material: DURAN™
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass (3.3) |
| No. per Pack | 10 |
| Socket Joint | NS 45/40 |
| Diameter (Metric) Outer | 85 mm |