Learn More
Corning™ Rotor Adapter for LSE™ Mini Microcentrifuge 6770-RT
Product Code. 17213564 Shop All Corning Products
Change view
Click to view available options
Capacity:
0.2 mL
0.25 mL
0.5 mL
Unit Size:
Pack of 8
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
17213564 0.25 mL Pack of 8
17203564 0.2 mL Pack of 8
17223564 0.5 mL Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17213564 Supplier Corning™ Supplier No. 6770RTA.25
Only null left
Add to basket
Add to basket
Corning™ Rotor Adapter comes in three sizes and is for use with Corning Mini Microcentrifuge 6770
Corning™ Rotor Adapter comes in three sizes and is for use with Corning Mini Microcentrifuge 6770
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Capacity 0.25 mL
For Use With (Equipment) Corning™ Mini Microcentrifuge 6770
No. per Pack 8/Pk.