Learn More
Welch ILMVAC™ Rotary Vane Pump
Product Code. p-6980064 Shop All Welch Ilmvac Products
Change view
Click to view available options
Flow Rate:
183/220 L/min.
96.7/110 L/min.
Maximum Pressure:
35 mbar
60 mbar
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Maximum Pressure unitSize
11969078 183/220 L/min. 35 mbar Each
10285789 96.7/110 L/min. 60 mbar Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11969078 Supplier Welch ILMVAC™ Supplier No. 322003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Withstands large amounts of water vapor in the vacuum pump’s inlet vapor stream. Welch ILMVAC™ Rotary Vane Pump is low maintenance, compact, and exceptionally quiet.
  • Two stage rotary vane pump
  • High water vapor tolerance (60 mbar for P6Z and 35 mbar for P12Z)
  • Max. leak rate of inlet valve 7x10-5 mbar l/sec
  • Integrated oil control glass
  • High temperature heat dissipation
  • Easy to service and clean
TRUSTED_SUSTAINABILITY
Specifications
Horsepower 550 w
Dimensions (L x W x H) 180 x 500 x 280 mm
Model P12Z
Weight (Metric) 27 kg
Flow Rate 183/220 L/min.
Frequency 50/60 Hz
Pump Type Rotary Vane Vacuum Pump
Applications Backing Pump
Maximum Pressure 35 mbar