Learn More
Hirschmann Instruments™ rotarus™ Volume 50 Peristaltic Pump
Click to view available options
Channels:
1 to 12 (Depending pump head)
Unit Size:
Each
Description
- Changeable pump heads, single and multichannel design
- Rapid tool-free changing of pump heads
- Automatic pump head and hose detection with RFID technology
- Control panel located directly on pump housing
- TFT touchscreen with user-friendly menu
- Calibratable
- Method memory
- Operation with 24 V DC
- USB and RS232 interface for PC link using interface (available as accessory)
- Pump heads from other manufacturers can be fitted without difficulty using an adapter
- Comprehensive accessory range
Specifications
Specifications
| Channels | 1 to 12 (Depending pump head) |
| Motor Type | Stepper Motor |
| Dimensions (L x W x H) | 205 x 100 x 220 mm |
| Weight (Metric) | 7.3 kg |
| Voltage | 100/240 VAC |
| Frequency | 50/60 Hz |