missing translation for 'onlineSavingsMsg'
Learn More

Rosuvastatin Calcium Salt, TRC

Product Code. 30419923
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419923 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419923 Supplier Toronto Research Chemicals Supplier No. TRCR70050050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rosuvastatin Calcium
CAS 147098-20-2
Purity Grade Notes HPLC
Molecular Formula 2 C22 H27 F N3 O6 S . Ca
Quantity 50 mg
Synonym 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, calcium salt (2:1), (3R,5S,6E)-, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, calcium salt (2:1), [S-[R*,S*-(E)]]-, Crestomed, Crestor, Fortius, Rostar, Rosuvastatin calcium, Rosuvastatin calcium salt, Rosuvastatin hemicalcium, Rozavel, S 4522, Suvikan, ZD 4522, calcium salt
InChI Formula InChI=1S/2C22H28FN3O6S.Ca/c2*1-13(2)20-18(10-9-16(27)11-17(28)12-19(29)30)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)33(4,31)32;/h2*5-10,13,16-17,27-28H,11-12H2,1-4H3,(H,29,30);/q;;+2/p-2/b2*10-9+;/t2*16-,17-;/m11./s1
SMILES [Ca+2].CC(C)c1nc(nc(c1\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c2ccc(F)cc2)N(C)S(=O)(=O)C.CC(C)c3nc(nc(c3\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c4ccc(F)cc4)N(C)S(=O)(=O)C
IUPAC Name calcium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
Molecular Weight (g/mol) 1001.14
Formula Weight 1000.2835
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.