missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rosoxacin Potassium Salt (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | Rosoxacin Potassium Salt |
| Molecular Formula | C17H13KN2O3 |
| Quantity | 10 mg |
| Synonym | 1-Ethyl-1,4-dihydro-4-oxo-7-(4-pyridinyl)-3-quinolinecarboxylic Acid Potassium Salt, Acrosoxacin Potassium Salt, Eracin Potassium Salt, Eradacil Potassium Salt, Eradacin Potassium Salt, NSC 146617 Potassium Salt, Roxadyl Potassium Salt, Win 35213 Potassium Salt, Winuron Potassium Salt, 1-Ethyl-4-oxo-7-(pyridin-4-yl)-1,4-dihydroquinoline-3-carboxylic Acid Potassium Salt |
| InChI Formula | InChI=1S/C17H14N2O3.K/c1-2-19-10-14(17(21)22)16(20)13-4-3-12(9-15(13)19)11-5-7-18-8-6-11;/h3-10H,2H2,1H3,(H,21,22);/q;+1/p-1 |
| SMILES | O=C(C1=CN(CC)C2=C(C=CC(C3=CC=NC=C3)=C2)C1=O)[O-].[K+] |
| IUPAC Name | potassium 1-ethyl-4-oxo-7-(pyridin-4-yl)-1,4-dihydroquinoline-3-carboxylate |
| Molecular Weight (g/mol) | 332.4 |
| Formula Weight | 332.06 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?