missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ropivacaine Hydrochloride Monohydrate, Mikromol™
Click to view available options
Quantity:
250 mg
missing translation for 'unitSize'
250mg
Description
Ropivacaine Hydrochloride Monohydrate
Specifications
Specifications
| Chemical Name or Material | Ropivacaine hydrochloride monohydrate |
| CAS | 132112-35-7 |
| Analyte or Component Names | Ropivacaine hydrochloride monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H26 N2 O . Cl H . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ropivacaine hydrochloride monohydrate, (-)-(2S)-N-(2,6-Dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride monohydrate |
| InChI Formula | InChI=1S/C17H26N2O.ClH.H2O/c1-4-11-19-12-6-5-10-15(19)17(20)18-16-13(2)8-7-9-14(16)3;;/h7-9,15H,4-6,10-12H2,1-3H3,(H,18,20);1H;1H2/t15-;;/m0../s1 |
| SMILES | O.Cl.CCCN1CCCC[C@H]1C(=O)Nc2c(C)cccc2C |
| IUPAC Name | (2S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide;hydrate;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction