missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ropivacaine hydrochloride monohydrate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ropivacaine hydrochloride monohydrate is a potent sodium channel blocker and blocks impulse conduction via reversible inhibition of sodium ion influx in nerve fibrese. Ropivacaine is also an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. Ropivacaine is widely used for regional anesthesia and neuropathic pain management in vivo.
Specifications
Specifications
| Chemical Name or Material | Ropivacaine hydrochloride monohydrate |
| CAS | 132112-35-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H29ClN2O2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 100 mg/mL (304.06 mM; Need ultrasonic) ∣H2O : 50 mg/mL (152.03 mM; Need ultrasonic) |
| SMILES | O=C([C@H]1N(CCC)CCCC1)NC2=C(C)C=CC=C2C.O.Cl |
| Molecular Weight (g/mol) | 328.88 |
| Formula Weight | 328.88 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction