missing translation for 'onlineSavingsMsg'
Learn More

Roluperidone, MedChemExpress

Product Code. 16460316
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460316

Brand: Medchem Express HY19469

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Roluperidone (CYR-101) is a novel cyclic amide derivative that has high equipotent affinities for 5-HT2A and sigma-2 receptors (Ki of 7.53 nM and 8.19 nM for 5-HT2A and sigma-2, respectively).

Roluperidone (CYR-101) is a novel cyclic amide derivative that has high equipotent affinities for 5-HT2A and sigma-2 receptors (Ki of 7.53 nM and 8.19 nM for 5-HT2A and sigma-2, respectively).

Specifications

Chemical Name or Material Roluperidone
CAS 359625-79-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H23FN2O2
Quantity 2 mg
Synonym CYR-101 MIN-101 MT-210
Solubility Information DMSO : 33.33 mg/mL (90.96 mM; Need ultrasonic)
SMILES O=C1N(CC2CCN(CC(C3=CC=C(F)C=C3)=O)CC2)CC4=C1C=CC=C4
Molecular Weight (g/mol) 366.43
Formula Weight 366.43
Percent Purity 98.08%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.