missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Roflumilast Impurity E, MedChemExpress
Description
Roflumilast Impurity E is the impurity of Roflumilast. Roflumilast(Daliresp) is a drug which acts as a selective and long-acting inhibitor of the enzyme PDE-4 with an IC50 value of 0.8 nM.
Specifications
Specifications
| Chemical Name or Material | Roflumilast Impurity E |
| CAS | 1391052-76-8 |
| Purity Grade Notes | Research |
| Molecular Formula | C13H8Cl2F2N2O3 |
| Quantity | 1 mg |
| SMILES | O=C(NC1=C(Cl)C=NC=C1Cl)C2=CC=C(OC(F)F)C(O)=C2 |
| Molecular Weight (g/mol) | 349.12 |
| Formula Weight | 349.12 |
| Grade | Research |
| For Use With (Application) | COVID-19-immunoregulation |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?