Learn More
Hamilton™ Robotics Low Bleed High Temperature Septa
Product Code. 10034202
Click to view available options
Diameter (Metric):
9.5 mm
12.7 mm
Unit Size:
Pack of 50
Product Code. 10034202
Brand: Hamilton™ 75833
Only null left
Specifications
Diameter (Metric) 9.5 mm
For Use With (Equipment) GC
Quantity 50/Pk.
Product Type Low Bleed High Temperature Septa