missing translation for 'onlineSavingsMsg'
Learn More

Ro 25-6981 Maleate, MedChemExpress

Product Code. p-200097623
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466386

Brand: Medchem Express HY13993A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ro 25-6981 Maleate is a potent, selective and activity-dependent NR2B subunit specific NMDA receptor antagonist. Ro 25-6981 Maleat shows anticonvulsant and anti-parkinsonian activity. Ro 25-6981 Maleate has the potential for the research of parkinson's disease (PD).

Ro 25-6981 Maleate is a potent, selective and activity-dependent NR2B subunit specific NMDA receptor antagonist. Ro 25-6981 Maleat shows anticonvulsant and anti-parkinsonian activity. Ro 25-6981 Maleate has the potential for the research of parkinson's disease (PD).

Specifications

Chemical Name or Material Ro 25-6981 Maleate
CAS 1312991-76-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H33NO6
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (219.52 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (18.29 mM; ultrasonic and warming and heat to 45°C)
SMILES OC1=CC=C([C@H](O)[C@@H](C)CN2CCC(CC3=CC=CC=C3)CC2)C=C1.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 455.54
Formula Weight 455.54
Percent Purity 97.48%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.