missing translation for 'onlineSavingsMsg'
Learn More

Ro 10-5824 dihydrochloride, MedChemExpress

Product Code. p-200096809
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16476067

Brand: Medchem Express HY101384A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist, with Ki of 5.2 nM.

Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist, with Ki of 5.2 nM.

Specifications

Chemical Name or Material Ro 10-5824 dihydrochloride
Physical Form Solid
CAS 189744-94-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H22Cl2N4
Solubility Information H2O : 100 mg/mL (283.05 mM; Need ultrasonic) ∣DMSO : 8.33 mg/mL (23.58 mM; Need ultrasonic)
SMILES NC1=NC(C)=NC=C1CN2CC=C(C3=CC=CC=C3)CC2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 353.29
Formula Weight 353.29
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.