missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH2B Rabbit anti-Human, Clone: RM433, Novus Biologicals™
Rabbit Monoclonal Antibody
209.00 EUR - 536.00 EUR
Specifications
| Antigen | RNASEH2B |
|---|---|
| Clone | RM433 |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:100-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18362443
|
Novus Biologicals
NBP3-09181-25UL |
25 μg |
209.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18329882
|
Novus Biologicals
NBP3-09181-100UL |
100 μg |
536.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNASEH2B Monoclonal specifically detects RNASEH2B in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| RNASEH2B | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:100-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, 50% Glycerol, 1% BSA | |
| 79621 | |
| IgG | |
| Protein A purified |
| RM433 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ribonuclease H2, subunit B | |
| A peptide corresponding to the C-terminus of RNASEH2B | |
| Primary | |
| Store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title