missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH2A Antibody (3G5-F5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00010535-M01
This item is not returnable.
View return policy
Description
RNASEH2A Monoclonal antibody specifically detects RNASEH2A in Human samples. It is validated for Western Blot, ELISA, ELISA, KnockDown
Specifications
| RNASEH2A | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| AGS4RNase H2 subunit A, Aicardi-Goutieres syndrome 4, Aicardi-Goutieres syndrome 4 protein, JUNB, ribonuclease H2 subunit A, ribonuclease H2, large subunit, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, ribonuclease HI, large subunit, RNase H(35), RNASEHIEC 3.1.26.4, RNHIARNase HI large subunit, RNHL | |
| RNASEH2A (AAH11748, 1 a.a. ~ 299 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDLSELERDNTGRCRLSSPVPAVCRKEPCVLGVDEAGRGPVLGPMVYAICYCPLPRLADLEALKVADSKTLLESERERLFAKMEDTDFVGWALDVLSPNLISTSMLGRVKYNLNSLSHDTATGLIQYALDQGVNVTQVFVDTVGMPETYQARLQQSFPGIEVTVKAKADALYPVVSAASICAKVARDQAVKKWQFVEKLQDLDTDYGSGYPNDPKTKAWLKEHVEPVFGFPQFVRFSWRTAQTILEKEAEDVIWEDSASENQEGLRKITSYFLNEGSQARPRSSHRYFLERGLESATSL | |
| 0.1 mg | |
| DNA replication Transcription Translation and Splicing | |
| 10535 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Sandwich ELISA, KnockDown | |
| 3G5-F5 | |
| Western Blot 1:500 | |
| AAH11748 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction