Learn More
RN-18, MedChemExpress
Product Code. 16493678
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16493678
Brand: Medchem Express HY102014
MedChemExpress RN-18 is a HIV-1 viral infectivity factor (HIV-1 Vif) inhibitor with an IC50 of 6 μM in nonpermissive H9 cells.
RN-18 is a HIV-1 viral infectivity factor (HIV-1 Vif) inhibitor with an IC50 of 6 μM in nonpermissive H9 cells.
Specifications
Chemical Name or Material RN-18
CAS 431980-38-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H16N2O4S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (262.87 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CC=C1OC)C2=CC=CC=C2SC3=CC=C([N+]([O-])=O)C=C3
Molecular Weight (g/mol) 380.42
Formula Weight 380.42
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus