missing translation for 'onlineSavingsMsg'
Learn More

RK-287107, MedChemExpress

Product Code. 16471367
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471367 50 mg 50mg
16447857 5 mg 5mg
16445138 10 mg 10mg
16415856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471367 Supplier Medchem Express Supplier No. HY123892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RK-287107 is a potent and specific tankyrase inhibitor with IC50s of 14.3 and 10.6 nM for tankyrase-1 and tankyrase-2, respectively. RK-287107 blocks colorectal cancer cell growth.

RK-287107 is a potent and specific tankyrase inhibitor with IC50s of 14.3 and 10.6 nM for tankyrase-1 and tankyrase-2, respectively. RK-287107 blocks colorectal cancer cell growth.

Specifications

Chemical Name or Material RK-287107
CAS 2171386-10-8
Color White
Purity Grade Notes Research
Molecular Formula C22H26F2N4O2
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (300.15 mM; Need ultrasonic)
SMILES O=C1NC(N2CCC3(CC2)CN(CCO)C4=C3C(F)=CC(F)=C4)=NC5=C1CCCC5
Molecular Weight (g/mol) 416.46
Formula Weight 416.46
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.