missing translation for 'onlineSavingsMsg'
Learn More

Rivastigmine-d4 Tartrate Salt, TRC

Product Code. 30411617
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411617 10 mg 10mg
30420511 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30411617 Supplier Toronto Research Chemicals Supplier No. TRCR54100410MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Rivastigmine-d4 Tartrate Salt
Molecular Formula C18H24D4N2O8
Quantity 10 mg
Synonym N-Ethyl-N-methylcarbamic Acid 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ester-d4 (2R,3R)-2,3-Dihydroxybutanedioate, (S)-Ethylmethylcarbamic Acid 3-[1-(Dimethylamino)ethyl]phenyl Ester-d4 [R-(R*,R*)]-2,3-Dihydroxybutanedioate
InChI Formula InChI=1S/C14H22N2O2.C4H6O6/c1-6-16(5)14(17)18-13-9-7-8-12(10-13)11(2)15(3)4;5-1(3(7)8)2(6)4(9)10/h7-11H,6H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t11-;1-,2-/m01/s1/i3D2,4D2;
SMILES CCN(C)C(OC1=CC([C@H](C)N(C([2H])([2H])[H])C([2H])([2H])[H])=CC=C1)=O.O[C@@H](C(O)=O)[C@H](C(O)=O)O
Molecular Weight (g/mol) 404.45
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.