missing translation for 'onlineSavingsMsg'
Learn More

Ritonavir Ethyl Carbamate, TRC

Product Code. 30422018
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422018 50 mg 50mg
30428164 5 mg 5mg
30409844 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422018 Supplier Toronto Research Chemicals Supplier No. TRCR53505050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ritonavir Ethyl Carbamate
Analyte or Component Names Ritonavir Ethyl Carbamate
Shipping Condition Room Temperature
Molecular Formula C31 H40 N4 O6 S
InChI Formula InChI=1S/C31H40N4O6S/c1-4-40-31(39)35-28(21(2)3)29(37)33-24(15-22-11-7-5-8-12-22)17-27(36)26(16-23-13-9-6-10-14-23)34-30(38)41-19-25-18-32-20-42-25/h5-14,18,20-21,24,26-28,36H,4,15-17,19H2,1-3H3,(H,33,37)(H,34,38)(H,35,39)/t24-,26-,27-,28-/m0/s1
Solution Type Neat
SMILES CCOC(=O)N[C@@H](C(C)C)C(=O)N[C@H](C[C@H](O)[C@H](Cc1ccccc1)NC(=O)OCc2cncs2)Cc3ccccc3
IUPAC Name 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-5-[[(2S)-2-(ethoxycarbonylamino)-3-methylbutanoyl]amino]-3-hydroxy-1,6-diphenylhexan-2-yl]carbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 596.737
Quantity 50 mg
Formula Weight 596.267
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.