missing translation for 'onlineSavingsMsg'
Learn More

RIPK1-IN-7, MedChemExpress

Product Code. 16495676
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495676

Brand: Medchem Express HY119933

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RIPK1-IN-7 is a potent and selective RIPK1 inhibitor with a Kd of 4 nM and an enzymatic IC50 of 11 nM. RIPK1-IN-7 exhibits excellent antimetastasis activity in the experimental B16 melanoma lung metastasis model.

RIPK1-IN-7 is a potent and selective RIPK1 inhibitor with a Kd of 4 nM and an enzymatic IC50 of 11 nM. RIPK1-IN-7 exhibits excellent antimetastasis activity in the experimental B16 melanoma lung metastasis model.

Specifications

Chemical Name or Material RIPK1-IN-7
CAS 2300982-44-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H22F3N5O2
Quantity 100 mg
Solubility Information DMSO : 62.5 mg/mL (129.81 mM; Need ultrasonic)
SMILES NC1=C2C(N(CC)C=C2C3=CC(CCN4C(CC5=CC=CC(OC(F)(F)F)=C5)=O)=C4C=C3)=NC=N1
Molecular Weight (g/mol) 481.47
Formula Weight 481.47
Percent Purity 98.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.