missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIOK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35219-20ul
This item is not returnable.
View return policy
Description
RIOK2 Polyclonal antibody specifically detects RIOK2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| RIOK2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 2.7.11.1, FLJ11159, RIO kinase 2, RIO kinase 2 (yeast), RIO2, serine/threonine-protein kinase RIO2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 253-552 of human RIOK2 (NP_060813.2).,, Sequence:, TSHPNAEWYFDRDVKCIKDFFMKRFSYESELFPTFKDIRREDTLDVEVSASGYTKEMQADDELLHPLGPDDKNIETKEGSEFSFSDGEVAEKAEVYGSENESERNCLEESEGCYCRSSGDPEQIKEDSLSEESADARSFEMTEFNQALEEIKGQVVENNSVTEFSEEKNRTENYNRQDGQRVQGGVPAGSDEYEDECPHLIALSSLNREFRPFRDEENVGAMNQYRTRTLSITSSGSAVSCSTIPPELVKQKVKRQLTKQQKSAVRRRLQKGEANIFTKQRRENMQNIKSSLEAASFWGE | |
| 20 μL | |
| Protein Kinase | |
| 55781 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction