missing translation for 'onlineSavingsMsg'
Learn More

Rimeporide, MedChemExpress

Product Code. 16440178
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16440178 5 mg 5mg
16451789 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16440178 Supplier Medchem Express Supplier No. HY19273

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rimeporide (EMD-87580) is a potent and selective inhibitor of the Na+/H+ exchanger (NHE-1).

Rimeporide (EMD-87580) is a potent and selective inhibitor of the Na+/H+ exchanger (NHE-1).

Specifications

Chemical Name or Material Rimeporide
CAS 187870-78-6
Color White
Purity Grade Notes Research
Molecular Formula C11H15N3O5S2
Quantity 5 mg
Synonym EMD-87580
Solubility Information DMSO : 20 mg/mL (59.99 mM; Need ultrasonic and warming)
SMILES O=C(NC(N)=N)C1=CC(S(=O)(C)=O)=C(S(=O)(C)=O)C=C1C
Molecular Weight (g/mol) 333.38
Formula Weight 333.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.