missing translation for 'onlineSavingsMsg'
Learn More

Ricinoleic Acid, MP Biomedicals™

Product Code. 11404772 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 11404772

Brand: MP Biomedicals 0219151491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ricinoleic Acid is used as a substrate for the synthesis of the conjugated linoleic acids.

  • Ricinoleic Acid is a naturally occurring 12-hydroxy fatty acid constituting about 90% of the fatty acids in castor oil.
  • Has the capacity to serve as a substrate for the synthesis of the conjugated linoleic acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-22-0
Molecular Formula C18H34O3
Molecular Weight (g/mol) 298.47
MDL Number MFCD00084840
InChI Key WBHHMMIMDMUBKC-QJWNTBNXSA-N
Synonym ricinoleic acid, ricinolic acid, ricinic acid, r,z-12-hydroxyoctadec-9-enoic acid, kyselina ricinolova, castor oil acid, 12-hydroxy-cis-9-octadecenoic acid, nouracid cs 80, 9-octadecenoic acid, 12-hydroxy-, r-z, kyselina 12-hydroxy-9-oktadecenova
PubChem CID 643684
ChEBI CHEBI:28592
IUPAC Name (9Z,12R)-12-hydroxyoctadec-9-enoic acid
SMILES CCCCCC[C@@H](O)C\C=C/CCCCCCCC(O)=O

Specifications

Melting Point 5.5°C
Color Colorless
Density 0.940 g/mL at 27.4°C/4°C
Boiling Point 245°C
Assay Percent Range >99%
Quantity 1 g
Solubility Information Water Solubility: 3460 mg/L at 25°C)
Formula Weight 298.5
Vapor Pressure 4.49X10-3 mm Hg at 25°C
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Ricinoleic Acid
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.