missing translation for 'onlineSavingsMsg'
Learn More

Ricinine, TRC

Product Code. 30133795
Change view
Click to view available options
Quantity:
1 mg
25 mg
100 mg
Unit Size:
100mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133795 100 mg 100mg
18093710 25 mg 25mg
18031110 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133795 Supplier Toronto Research Chemicals Supplier No. TRCR495550100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-40-3
Molecular Formula C8 H8 N2 O2
Molecular Weight (g/mol) 164.16
Synonym 3-Pyridinecarbonitrile, 1,2-dihydro-4-methoxy-1-methyl-2-oxo-, Nicotinonitrile, 1,2-dihydro-4-methoxy-1-methyl-2-oxo- (7CI,8CI), Ricinine (6CI), 1,2-Dihydro-4-methoxy-1-methyl-2-oxo-3-pyridinecarbonitrile, 1,2-Dihydro-4-methoxy-1-methyl-2-oxonicotinonitrile, NSC 409913, NSC 642604, Recinine, Ricinin
IUPAC Name 4-methoxy-1-methyl-2-oxopyridine-3-carbonitrile
SMILES COC1=C(C#N)C(=O)N(C)C=C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H8N2O2/c1-10-4-3-7(12-2)6(5-9)8(10)11/h3-4H,1-2H3
Formula Weight 164.0586
Percent Purity >95%
Chemical Name or Material Ricinine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.