Learn More
Epredia™ Richard-Allan Scientific™ Mucicarmine, Tartrazine Stain Solution
Product Code. 12573657 Shop All Epredia Products
Click to view available options
Type:
Mucicarmine Stock Solution
Tartrazine Solution
Unit Size:
500mL
Product Code. 12573657
Brand: Epredia™ 88037
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Identify primary tumor sites with the aid of Epredia™ Richard-Allan Scientific™ Mucicarmine
  • Aids in distinguishing mucin-negative undifferentiated squamous cell lesions from mucin-positive adenocarcinomas
  • Mucin is stained dark rose and the nuclei are black
  • Capsule of Cryptococcus (fungus) is stained deep rose
  • Tartrazine solution is used as a yellow cytoplasmic stain
TRUSTED_SUSTAINABILITY
Specifications
Type Mucicarmine Stock Solution