Learn More
Epredia™ Richard-Allan Scientific™ Mucicarmine, Tartrazine Stain Solution
Shop All Epredia Products
Click to view available options
Type:
Mucicarmine Stock Solution
Tartrazine Solution
Unit Size:
500mL
Description
- Aids in distinguishing mucin-negative undifferentiated squamous cell lesions from mucin-positive adenocarcinomas
- Mucin is stained dark rose and the nuclei are black
- Capsule of Cryptococcus (fungus) is stained deep rose
- Tartrazine solution is used as a yellow cytoplasmic stain
Specifications
Specifications
| Type | Tartrazine Solution |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur