Learn More
Epredia™ Richard-Allan Scientific™ Cover Glass
Product Code. 13292933 Shop All Epredia Products
Change view
Click to view available options
Thickness (Metric):
#1 (0.13 to 0.16 mm)
#1.5 (0.16 to 0.19 mm)
Length (Metric):
22 mm
24 mm
Width (Metric):
22 mm
50 mm
60 mm
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Length (Metric) Width (Metric) unitSize
13292933 #1.5 (0.16 to 0.19 mm) 22 mm 22 mm Each
15504573 #1 (0.13 to 0.16 mm) 24 mm 50 mm Pack of 10
13202943 #1.5 (0.16 to 0.19 mm) 24 mm 60 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13292933 Supplier Epredia™ Supplier No. 152222
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reduces defect costs that range from wasted resources to lost time with a consistent, reliable workflow, which includes quality cover glass.
Ordering Information:
One oz. pack
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 22 mm
Width (Metric) 22 mm
Thickness (Metric) #1.5 (0.16 to 0.19 mm)
Dimensions (L x W) 22 x 22 mm
Description Thickness no.1.5; 22 x 22mm