Learn More
Ribociclib succinate, MedChemExpress
Product Code. 16489367
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489367 5 mg 5mg
16462897 50 mg 50mg
16446936 100 mg 100mg
16446828 10 mg 10mg
16401206 200 mg 200mg
16461786 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16489367 Supplier Medchem Express Supplier No. HY15777B
MedChemExpress Ribociclib succinate (LEE011 succinate) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex.
Ribociclib succinate (LEE011 succinate) is a highly specific CDK4/6 inhibitor with IC50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex.
Specifications
Chemical Name or Material Ribociclib succinate
CAS 1374639-75-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H36N8O5
Quantity 5 mg
Synonym LEE011 succinate
Solubility Information DMSO : 41.67 mg/mL (75.40 mM; Need ultrasonic) ∣H2O : 4.17 mg/mL (7.55 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N(C)C)C(N1C2CCCC2)=CC(C1=N3)=CN=C3NC(N=C4)=CC=C4N5CCNCC5.OC(CCC(O)=O)=O
Molecular Weight (g/mol) 552.63
Formula Weight 552.63
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease