missing translation for 'onlineSavingsMsg'
Learn More

Rho-Kinase-IN-1, MedChemExpress

Product Code. p-200100773
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16445637 5 mg 5mg
16422898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16445637 Supplier Medchem Express Supplier No. HY100270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rho-Kinase-IN-1 is a Rho kinase (ROCK) inhibitor (Ki values of 30.5 and 3.9 nM for ROCK1 and ROCK2, respectively) extracted from US20090325960A1, compound 1.008.

Rho-Kinase-IN-1 is a Rho kinase (ROCK) inhibitor (Ki values of 30.5 and 3.9 nM for ROCK1 and ROCK2, respectively) extracted from US20090325960A1, compound 1.008.

Specifications

Chemical Name or Material Rho-Kinase-IN-1
CAS 1035094-83-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H24N4S
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (141.84 mM; ultrasonic and warming and heat to 60°C)
SMILES CSC1=CC=C(CN2CC(NC3=CC4=C(NN=C4)C=C3)CCC2)C=C1
Molecular Weight (g/mol) 352.5
Formula Weight 352.5
Percent Purity 99.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.