Learn More
RGX-104 hydrochloride, MedChemExpress
Product Code. 16477197
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. 16477197
Brand: Medchem Express HY111498
MedChemExpress RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Specifications
Chemical Name or Material RGX-104 hydrochloride
CAS 610318-03-1
Color White
Purity Grade Notes Research
Molecular Formula C34H34Cl2F3NO3
Quantity 5 mg
Solubility Information DMSO : 150 mg/mL (237.14 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1.Cl
Molecular Weight (g/mol) 632.54
Formula Weight 632.54
Percent Purity 99.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation