Learn More
RGX-104 hydrochloride, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Description
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Specifications
Specifications
| Chemical Name or Material | RGX-104 hydrochloride |
| CAS | 610318-03-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C34H34Cl2F3NO3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 150 mg/mL (237.14 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1.Cl |
| Molecular Weight (g/mol) | 632.54 |
| Formula Weight | 632.54 |
| Show More |