missing translation for 'onlineSavingsMsg'
Learn More

RG7112, MedChemExpress

Product Code. 16407037
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16407037

Brand: Medchem Express HY10959

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RG7112 is a potent, selective, first clinical, orally active and blood-brain barrier crossed MDM2-p53 inhibitor, with an IC50 of 18 nM and a KD of 11 nM for binding to MDM2.

RG7112 is a potent, selective, first clinical, orally active and blood-brain barrier crossed MDM2-p53 inhibitor, with an IC50 of 18 nM and a KD of 11 nM for binding to MDM2.

Specifications

Chemical Name or Material RG7112
CAS 939981-39-2
Color White
Purity Grade Notes Research
Molecular Formula C38H48Cl2N4O4S
Quantity 5 mg
Synonym RO5045337
Solubility Information DMSO : 200 mg/mL (274.81 mM; Need ultrasonic)
SMILES C[C@@]1([C@](C)(N(C(C2=CC=C(C=C2OCC)C(C)(C)C)=N1)C(N3CCN(CC3)CCCS(=O)(C)=O)=O)C4=CC=C(C=C4)Cl)C5=CC=C(C=C5)Cl
Molecular Weight (g/mol) 727.78
Formula Weight 727.78
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.