Learn More
Thermo Scientific™ Reverse Osmosis Membranes
Product Code. 11830820 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Reverse Osmosis Membrane
Reverse Osmosis Membrane (pretreated), 3 L/hr.
Reverse Osmosis Membrane (pretreated), 6 L/hr.
For Use With (Equipment):
LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20
LabTower EDI 30, LabTower RO 40/60, LabTower TII 40/60, Pacific RO 40, Pacific TII 40
ROpure Infinity Systems
Smart2Pure
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) unitSize
11830820 Reverse Osmosis Membrane ROpure Infinity Systems Each
11377161 Reverse Osmosis Membrane LabTower EDI 30, LabTower RO 40/60, LabTower TII 40/60, Pacific RO 40, Pacific TII 40 Each
11377131 Reverse Osmosis Membrane (pretreated), 3 L/hr. Smart2Pure Each
11327141 Reverse Osmosis Membrane (pretreated), 6 L/hr. Smart2Pure Each
11337161 Reverse Osmosis Membrane LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11830820 Supplier Thermo Scientific™ Supplier No. D9006
Only null left
Add to basket
Add to basket
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Use Membranes with the Following Barnstead Systems:
09.2003:
  • Reverse osmosis membrane with integrated pretreatment, 3L/hr for Smart2Pure™ systems
09.2006:
  • Reverse osmosis membrane with integrated pretreatment, 6L/hr for Smart2Pure systems
22.0046:
  • LabTower™ EDI 15 requires two of these RO membranes
  • LabTower™ RO 20 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • LabTower™ TII 20 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • Pacific™ RO 3, 7, 12 require one of these RO membranes
  • Pacific RO 20 requires two of these RO membranes
  • Pacific™ TII 3, 7, 12 require one of these RO membranes
  • Pacific TII 20 requires two of these RO membranes
22.0087:
  • LabTower™ EDI 30 requires two of these RO membranes
  • LabTower™ RO 40 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • LabTower TII 40 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • Pacific RO 40 requires two of these RO membranes
  • Pacific TII 40 requires two of these RO membranes
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ROpure Infinity Systems
Type Reverse Osmosis Membrane