Learn More
RETSCH Analytical Sieve Shaker AS 200 Digit Test Sieve Lid
Features improved handling and service life for the sieves
Brand:  RETSCH 031070448
292.60 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 1.00000kg
Product Code. 11763369
  • 308.00 EUR / Each
Estimated Shipment: 01-08-2024
Specifications
Lid for test sieves 203 mm (8in.) (miniscule) x 1in.
Air Jet Sieving Machine AS 200 Jet