Learn More
Brady™ Retro Reflective All-weather Labels
Click to view available options
Dimensions (LxW):
12.70 mm (W) x 6.10 m (L)
19.05 mm (W) x 6.10 m (L)
25.40 mm (W) x 6.10 m (L)
38.10 mm (W) x 6.10 m (L)
Color:
White
Yellow
For Use With (Equipment):
BMP41, BMP51, BMP53, M511, M410, M510
BMP51, BMP53, M511, M510
Unit Size:
Each
Description
- Ideal for safety applications such as on personnel, fire suppression equipment, and first aid cabinets or anywhere extra visibility is required
- Resistant to abrasions, weather, solvents and extreme temperatures
- Pressure-sensitive acrylic adhesive withstands prolonged exposure to tough conditions
- Able to withstand outdoor conditions for 4 to 6 years on average
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | 2011/65/EU, 2015/863 |
| Adhesive Type | Permanent Acrylic |
| Coating | None |
| Color | Yellow |
| Finish | Gloss |
| Format | Continuous |
| Material | Plastic |
| Printable Sides | Top |
| Print Technology | Thermal Transfer |
| Show More |