Learn More
Motic™ Reticle Holder for 18mm Eyepiece
Click to view available options
For Use With (Equipment):
Eyepiece reticle, 18mm
Unit Size:
Each
Description
- Holds reticle for 18mm eyepiece..
- Protects from dust.
- Protects from damage.
Specifications
Specifications
| Type | Reticle Holder |
| For Use With (Equipment) | Eyepiece reticle, 18mm |