missing translation for 'onlineSavingsMsg'
Learn More

Retene, 98%, MP Biomedicals

Product Code. 11492676 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492676 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11492676 Supplier MP Biomedicals Supplier No. 0521789301

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 483-65-8
Molecular Formula C18H18
MDL Number MFCD00016358
Quantity 1 g
Synonym retene, reten, 1-methyl-7-isopropylphenanthrene, 7-isopropyl-1-methylphenanthrene, phenanthrene, 1-methyl-7-1-methylethyl, 1-methyl-7-1-methylethyl phenanthrene, unii-0w2d2e1p9q, phenanthrene, 7-isopropyl-1-methyl, ccris 3180, 7-isopropyl-1-methylphenanthren
InChI Key NXLOLUFNDSBYTP-UHFFFAOYSA-N
SMILES CC(C)C1=CC=C2C(C=CC3=C(C)C=CC=C23)=C1
IUPAC Name 1-methyl-7-(propan-2-yl)phenanthrene
Molecular Weight (g/mol) 234.34
PubChem CID 10222
Formula Weight 234.32
Chemical Name or Material Retene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.