missing translation for 'onlineSavingsMsg'
Learn More

Reserpine hydrochloride, MedChemExpress

Product Code. 16409096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16409096 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16409096 Supplier Medchem Express Supplier No. HYN0480A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Reserpine hydrochloride is an inhibitor of the vesicular monoamine transporter 2 (VMAT2).

Reserpine hydrochloride is an inhibitor of the vesicular monoamine transporter 2 (VMAT2).

Specifications

Chemical Name or Material Reserpine hydrochloride
CAS 16994-56-2
Color White
Purity Grade Notes Research
Molecular Formula C33H41ClN2O9
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (77.50 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C([C@H]([C@@H](OC)[C@H](OC(C1=CC(OC)=C(OC)C(OC)=C1)=O)C[C@]2([H])CN3CC4)[C@@]2([H])C[C@]3([H])C5=C4C(C=CC(OC)=C6)=C6N5)OC.Cl
Molecular Weight (g/mol) 645.14
Formula Weight 645.14
Percent Purity 99.65%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.