missing translation for 'onlineSavingsMsg'
Learn More

Resatorvid, MedChemExpress

Product Code. p-200096222
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16417127

Brand: Medchem Express HY11109

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Resatorvid (TAK-242) is a selective Toll-like receptor 4 (TLR4) inhibitor. Resatorvid inhibits NO, TNF-α and IL-6 production with IC50s of 1.8 nM, 1.9 nM and 1.3 nM, respectively. Resatorvid downregulates expression of TLR4 downstream signaling molecules MyD88 and TRIF. Resatorvid inhibits autophagy and plays pivotal role in various inflammatory diseases.

Resatorvid (TAK-242) is a selective Toll-like receptor 4 (TLR4) inhibitor. Resatorvid inhibits NO, TNF-α and IL-6 production with IC50s of 1.8 nM, 1.9 nM and 1.3 nM, respectively. Resatorvid downregulates expression of TLR4 downstream signaling molecules MyD88 and TRIF. Resatorvid inhibits autophagy and plays pivotal role in various inflammatory diseases.

Specifications

Chemical Name or Material Resatorvid
CAS 243984-11-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H17ClFNO4S
Quantity 5 mg
Synonym TAK-242 CLI-095
Solubility Information DMSO : 100 mg/mL (276.38 mM; Need ultrasonic)
SMILES O=S([C@H]1C(C(OCC)=O)=CCCC1)(NC2=CC=C(C=C2Cl)F)=O
Molecular Weight (g/mol) 361.82
Formula Weight 361.82
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.