Learn More
BRAND™ Replacement PFA Screw Cap
Product Code. p-7164791 Shop All Brand Products
Click to view available options
Closure Size:
GL 18
GL 25
Unit Size:
Each
Product Code. 10047362
Brand: BRAND™ 129250
Only null left
Add to basket
Add to basket
Replacement PFA Screw Cap suitable for use with a range of volumetric flasks.
  • Constructed of PFA
  • For use with volumetric flasks
  • Replacement
TRUSTED_SUSTAINABILITY
Specifications
Material PFA
For Use With (Equipment) Volumetric Flask
Height (Metric) 2 cm
Length (Metric) 2.4 cm
Product Type Screw cap
Width (Metric) 2 cm
Closure Size GL 18