Learn More
Hamilton™ Replacement Needles for Waters U6K HPLC Injection Valve Syringes
Click to view available options
Needle Point Style:
Removable Square/Blunt
Product Type:
Large Hub Replacement Needle
Small Hub Replacement Needle
Unit Size:
Pack of 6
Description
Needles are available as a 25s gauge with a small hub for syringes 2.5-100οL in volume, a large hub for syringes 250οL and larger, and metal hub needles for TLL syringes.
Six needles per pack.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Needle Gauge | 25s G |
| Length (English) Needle | 1.97 in. |
| Product Type | Large Hub Replacement Needle |
| Needle Point Style | Removable Square/Blunt |
| For Use With (Equipment) | Waters U6K HPLC Injection Valve Syringes |