Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Product Code. 12759290
Click to view available options
Description:
Polyvalent, Phase 2
Polyvalent, Phases 1 and 2
Salmonella H (1, 2)
Salmonella H (1, 5)
Salmonella H (1, 6)
Salmonella H (1, 7)
Salmonella H (E) Polyvalent
Salmonella H (G) Polyvalent
Salmonella H (a)
Salmonella H (b)
Salmonella H (d)
Salmonella H (enx)
Salmonella H (f, g)
Salmonella H (gm)
Salmonella H (i)
Unit Size:
2mL
Each
Product Code. 12759290
Brand: Thermo Scientific™ R30161201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Control Sets Positive
Description Salmonella H (gm)
CE Marker Yes
Detectable Analytes Salmonella H Antigen
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella