Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis
Product Code. 13245589
Click to view available options
Description:
Group A
Group B
Group C
Group D
Group W135
Unit Size:
2mL
Product Code. 13245589
Brand: Thermo Scientific™ R30166801
Used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description Group A
CE Marker Yes
Detectable Analytes N. meningitidis
Type Neisseria Meningitidis Test Kit
Organism Group Group A
Target Organism Class Neisseria Meningitidis