Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
382.00 EUR - 420.00 EUR
Product Code | Brand | Description | Detectable Analytes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Detectable Analytes | Price | Quantity & Availability | ||||
12627459
|
Thermo Scientific™
R30166001 |
Agglutinating Sera, Haemophilus influenzae Type A | H. influenzae |
419.00 EUR
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12653528
|
Thermo Scientific™
R30166401 |
Agglutinating Sera, Haemophilus influenzae Type E | H. influenzae |
382.00 EUR
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12663528
|
Thermo Scientific™
R30166501 |
Agglutinating Sera, Haemophilus influenzae Type F | H. influenzae |
382.00 EUR
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11464069
|
Thermo Scientific™
R30166101 |
Agglutinating Sera, Haemophilus influenzae Type B | H. influenzae |
420.00 EUR
2mL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12637459
|
Thermo Scientific™
R30166201 |
Agglutinating Sera, Haemophilus influenzae Type C | H. influenzae |
382.00 EUR
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12643528
|
Thermo Scientific™
R30166301 |
Agglutinating Sera, Haemophilus influenzae Type D | H. influenzae |
382.00 EUR
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Specifications
H. influenzae | |
Agglutinating Sera |